Learn More
Matrix Scientific (2R,3R,4S,5S,6S)-2-Bromo-6-(methoxycarbonyl)-tetrahydro-2H-pyran-3,4,5-triyl triacetate, 21085-72-3, MFCD00061613, 1g
Catalog No. 50691987
Encompass_Preferred
Molecular Formula C13H17BrO9, Purity 95+%, Molecular Weight 397.18, Melting Point 106-107°
Catalog No. 50-691-987 Supplier Matrix Scientific Supplier No. 0986201G
Molecular Formula C13H17BrO9, Purity 95+%, Molecular Weight 397.18, Melting Point 106-107°