Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2R 3R-E1R 10mg | 1400888-63-2 | 232.28 | C13H16N2O2 | 10 MG
SDP

Catalog No. 5000307950
Encompass_Preferred

(2R,3R)-E1R is the (2R,3R) enantiomer of E1R and a sigma-1 receptor positive allosteric modulator (Sig1R PAM) supplied for research use. It is intended for in vitro and preclinical studies investigating cognition and memory disorders and is not for human or clinical use.

  • Sigma-1 receptor positive allosteric modulator for cognition and memory research.
  • High chemical purity: 98.8% and enantiomeric excess 99.6%.
  • Chemical formula C13H16N2O2, molecular weight 232.28.
  • Available in small milligram packages (for example, 10 MG).
  • Storage: powder -20°C (up to 3 years) or 4°C (up to 2 years); in solvent -80°C (up to 6 months).

Catalog No. 50-003-07950 Supplier Medchemexpress LLC Supplier No. HY116463C10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(2R,3R)-E1R is the (2R,3R) enantiomer of E1R and a sigma-1 receptor positive allosteric modulator (Sig1R PAM) supplied for research use. It is intended for in vitro and preclinical studies investigating cognition and memory disorders and is not for human or clinical use.

  • Sigma-1 receptor positive allosteric modulator for cognition and memory research.
  • High chemical purity: 98.8% and enantiomeric excess 99.6%.
  • Chemical formula C13H16N2O2, molecular weight 232.28.
  • Available in small milligram packages (for example, 10 MG).
  • Storage: powder -20°C (up to 3 years) or 4°C (up to 2 years); in solvent -80°C (up to 6 months).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.