Learn More
Medchemexpress LLC HY-116463C 5mg Medchemexpress, (2R,3R)-E1R CAS:1400888-63-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY116463C5MG
Encompass_Preferred
Medchemexpress, HY-116463C 5mg (2R,3R)-E1R CAS:1400888-63-2 (2R,3R)-E1R (Compound 2b) is an enantiomer of E1R. (2R,3R)-E1R is a sigma-1 receptor positive allosteric modulator (Sig1R PAM) for the treatment of cognition/memory disorders. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9425