Learn More
Medchemexpress LLC (2R,3R)-Firazorexton | 2692692-00-3 | 99.9% | 470.51 | 1 ML
SDP
Catalog No. 5000351333
Encompass_Preferred
(2R,3R)-Firazorexton ((2R,3R)-TAK-994 free base) is an isomer of Firazorexton. It is an orally active, brain-penetrating orexin type 2 receptor (OX2R) agonist that may improve narcolepsy-like symptoms.
  • Orally active
  • Brain-penetrating
  • Orexin type 2 receptor (OX2R) agonist
  • May improve narcolepsy-like symptoms
  • High purity (99.85%)
Catalog No. 50-003-51333 Supplier Medchemexpress LLC Supplier No. HY137440B10MM/1ML
(2R,3R)-Firazorexton ((2R,3R)-TAK-994 free base) is an isomer of Firazorexton. It is an orally active, brain-penetrating orexin type 2 receptor (OX2R) agonist that may improve narcolepsy-like symptoms.
  • Orally active
  • Brain-penetrating
  • Orexin type 2 receptor (OX2R) agonist
  • May improve narcolepsy-like symptoms
  • High purity (99.85%)