Learn More
Medchemexpress LLC (2R,3R)-Firazorexton | 2692692-00-3 | 99.9% | 470.51 g/mol | C22H25F3N2O4S | 5 MG
SDP
Catalog No. 5000222472
Encompass_Preferred
(2R,3R)-Firazorexton is an orally active, brain-penetrating orexin type 2 receptor (OX2R) agonist supplied for research use. It is offered as a solid powder and as a DMSO solution for in vitro and preclinical pharmacology studies addressing orexin signaling and narcolepsy-like models.
  • Orally active OX2R agonist for pharmacology research.
  • Provided as solid powder and 10 mM DMSO solution.
  • High purity (~99.85%) suitable for research assays.
  • Soluble in DMSO (≈200 mg/mL); ultrasonic assistance may be required.
  • Store powder at -20°C; store in solvent at -80°C for extended stability.
Catalog No. 50-002-22472 Supplier Medchemexpress LLC Supplier No. HY137440B5MG
(2R,3R)-Firazorexton is an orally active, brain-penetrating orexin type 2 receptor (OX2R) agonist supplied for research use. It is offered as a solid powder and as a DMSO solution for in vitro and preclinical pharmacology studies addressing orexin signaling and narcolepsy-like models.
  • Orally active OX2R agonist for pharmacology research.
  • Provided as solid powder and 10 mM DMSO solution.
  • High purity (~99.85%) suitable for research assays.
  • Soluble in DMSO (≈200 mg/mL); ultrasonic assistance may be required.
  • Store powder at -20°C; store in solvent at -80°C for extended stability.