Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (2R,3R)-GSK973 | 00-00-0 | >98.0% | 410.44 | C23H23FN2O4 | 5 MG
SDP

Catalog No. 5000222424
Encompass_Preferred

(2R,3R)-GSK973 is the (2R,3R) stereoisomer of GSK973, a selective, orally bioavailable inhibitor of the BD2 (second bromodomain) domains of the BET family. Supplied as a powder for research use, it is intended for biochemical and cellular assays probing bromodomain-mediated epigenetic regulation. The compound is provided in small milligram packages and has defined storage conditions to preserve stability.

  • Selective inhibitor of BD2 bromodomains.
  • Orally bioavailable in in vivo evaluations.
  • Supplied as a powder for laboratory use.
  • Molecular formula C23H23FN2O4.
  • Molecular weight 410.44.
  • Powder storage: -20°C, 3 years; in solvent: -80°C for 6 months or -20°C for 1 month.
  • Available package sizes: 1 mg, 5 mg, 10 mg.

Catalog No. 50-002-22424 Supplier Medchemexpress LLC Supplier No. HY138563A5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(2R,3R)-GSK973 is the (2R,3R) stereoisomer of GSK973, a selective, orally bioavailable inhibitor of the BD2 (second bromodomain) domains of the BET family. Supplied as a powder for research use, it is intended for biochemical and cellular assays probing bromodomain-mediated epigenetic regulation. The compound is provided in small milligram packages and has defined storage conditions to preserve stability.

  • Selective inhibitor of BD2 bromodomains.
  • Orally bioavailable in in vivo evaluations.
  • Supplied as a powder for laboratory use.
  • Molecular formula C23H23FN2O4.
  • Molecular weight 410.44.
  • Powder storage: -20°C, 3 years; in solvent: -80°C for 6 months or -20°C for 1 month.
  • Available package sizes: 1 mg, 5 mg, 10 mg.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.