Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(2R,3S)-1-Carboxy-4-pentyl-2,3-dihydroxycyclohexa-4,6-diene Potassium Salt 97%, Thermo Scientific™

Catalog No. AC271772500
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC271772500 250 mg
AC271770010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC271772500 Supplier Thermo Scientific Chemicals Supplier No. 271772500

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 205639-93-6
Molecular Formula C12H17KO4
Molecular Weight (g/mol) 264.362
InChI Key BBLPZPQHEWQLGM-VZXYPILPSA-M
Synonym potassium 5s,6r-5,6-dihydroxy-4-pentylcyclohexa-1,3-dienecarboxylate, 2r,3s-1-carboxy-4-pentyl-2,3-dihydroxycyclohexa-4,6-diene potassium salt
PubChem CID 23666349
IUPAC Name potassium;(5S,6R)-5,6-dihydroxy-4-pentylcyclohexa-1,3-diene-1-carboxylate
SMILES CCCCCC1=CC=C(C(C1O)O)C(=O)[O-].[K+]

Specifications

CAS Min % 96.0
CAS Max % 100.0
Quantity 250 mg
Formula Weight 264.35
Chemical Name or Material (2R,3S)-1-Carboxy-4-pentyl-2,3-dihydroxycyclohexa-4,6-diene potassium salt, 97%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.