Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (2R,3S,4E)-2-(methylamino)-5-(4-pentylphenyl)pent-4-ene-1,3-diol | 1072443-89-0 | >98.0% | 277.4 g/mol | C17H27NO2 | 100 MG
SDP

Catalog No. 5000228864
Encompass_Preferred

SK1-I (BML-258) is an isozyme-selective sphingosine kinase 1 (SPHK1) inhibitor used as a research tool to probe SPHK1-related signaling, autophagy, and cancer biology. It is provided as a solid for laboratory research use only and is commonly supplied in 100 mg packages.

  • Isozyme-selective SPHK1 inhibitor with reported Ki ≈ 10 μM.
  • Enhances autophagy and demonstrates antitumor activity in preclinical studies.
  • CAS RN 1072443-89-0.
  • Molecular formula C17H27NO2; molecular weight 277.4 g/mol (free base).
  • Purity greater than 98.0% by HPLC when specified.
  • Supplied as a solid; typical package 100 MG.

Catalog No. 50-002-28864 Supplier Medchemexpress LLC Supplier No. HY119016100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

SK1-I (BML-258) is an isozyme-selective sphingosine kinase 1 (SPHK1) inhibitor used as a research tool to probe SPHK1-related signaling, autophagy, and cancer biology. It is provided as a solid for laboratory research use only and is commonly supplied in 100 mg packages.

  • Isozyme-selective SPHK1 inhibitor with reported Ki ≈ 10 μM.
  • Enhances autophagy and demonstrates antitumor activity in preclinical studies.
  • CAS RN 1072443-89-0.
  • Molecular formula C17H27NO2; molecular weight 277.4 g/mol (free base).
  • Purity greater than 98.0% by HPLC when specified.
  • Supplied as a solid; typical package 100 MG.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.