Learn More
Medchemexpress LLC (2R,3S,5R)-2-(hydroxymethyl)-5-(6-methyl-9H-purin-9-yl)tetrahydrofuran-3-ol | 16006-64-7 | MFCD29059892 | 95.0% | 249.27 g/mol | C12H15N3O3 | 5 MG
SDP
Catalog No. 5000281047
Encompass_Preferred
9-(2-Deoxy-beta-D-ribofuranosyl)-6-methylpurine is a purine nucleoside analogue used as a research reagent and reference compound in biochemical and anticancer studies. It functions as a nucleoside antimetabolite that can inhibit DNA synthesis and induce apoptosis. The compound is supplied as an off-white to light yellow solid and is characterized by CAS number 16006-64-7, molecular formula C12H15N3O3, and molecular weight 249.27 g/mol.
  • Purity 95.0%.
  • Available in small quantities, including 5 mg, for screening and reference use.
  • Analytical documentation available: COA, 1H NMR, LCMS, and SDS.
  • Suitable for studies of nucleoside metabolism and anticancer mechanisms.
  • Solid form, easy to handle and store.
Catalog No. 50-002-81047 Supplier Medchemexpress LLC Supplier No. HYW3944325MG
9-(2-Deoxy-beta-D-ribofuranosyl)-6-methylpurine is a purine nucleoside analogue used as a research reagent and reference compound in biochemical and anticancer studies. It functions as a nucleoside antimetabolite that can inhibit DNA synthesis and induce apoptosis. The compound is supplied as an off-white to light yellow solid and is characterized by CAS number 16006-64-7, molecular formula C12H15N3O3, and molecular weight 249.27 g/mol.
  • Purity 95.0%.
  • Available in small quantities, including 5 mg, for screening and reference use.
  • Analytical documentation available: COA, 1H NMR, LCMS, and SDS.
  • Suitable for studies of nucleoside metabolism and anticancer mechanisms.
  • Solid form, easy to handle and store.