Learn More
Medchemexpress LLC (2R,3S)-Azelaprag | 2049979-31-7 | 99.1% | 523.61 | 25 MG
SDP
Catalog No. 5000395293
Encompass_Preferred
(2R,3S)-Azelaprag ((2R,3S)-AMG 986) is a chemical compound acting as a precursor of an apelin receptor agonist (EC50=12 nM) and an isomer of Azelaprag. This compound is suitable for various laboratory research applications, including those involving GPCR/G Protein studies and apelin receptor research.
  • Functions as a precursor of an apelin receptor agonist
  • Identified as an isomer of Azelaprag
  • Used as an inhibitor in research
  • Applicable in GPCR/G protein studies
  • Relevant for apelin receptor (APJ) research
Catalog No. 50-003-95293 Supplier Medchemexpress LLC Supplier No. 25MGHY167712
(2R,3S)-Azelaprag ((2R,3S)-AMG 986) is a chemical compound acting as a precursor of an apelin receptor agonist (EC50=12 nM) and an isomer of Azelaprag. This compound is suitable for various laboratory research applications, including those involving GPCR/G Protein studies and apelin receptor research.
  • Functions as a precursor of an apelin receptor agonist
  • Identified as an isomer of Azelaprag
  • Used as an inhibitor in research
  • Applicable in GPCR/G protein studies
  • Relevant for apelin receptor (APJ) research