Learn More
Medchemexpress LLC (2R,3S)-Azelaprag | 2049979-31-7 | 99.1% | 523.61 g·mol⁻¹ | C25H29N7O4S | 10 MG
SDP
Catalog No. 5000289335
Encompass_Preferred
(2R,3S)-Azelaprag is a research chemical described as a precursor to an apelin receptor agonist and an isomer of Azelaprag (AMG 986). It exhibits activity at the apelin receptor (EC50 ≈ 12 nM) and is supplied as a high-purity solid for laboratory research.
  • Precursor to apelin receptor agonists with reported EC50 ≈ 12 nM.
  • High purity solid (99.1%) suitable for analytical and preclinical studies.
  • Molecular formula C25H29N7O4S; molecular weight 523.61 g·mol⁻¹.
  • Available in small-scale milligram packages and as a DMSO solution for screening.
  • Intended for laboratory research and experimental use only.
Catalog No. 50-002-89335 Supplier Medchemexpress LLC Supplier No. HY16771210MG
(2R,3S)-Azelaprag is a research chemical described as a precursor to an apelin receptor agonist and an isomer of Azelaprag (AMG 986). It exhibits activity at the apelin receptor (EC50 ≈ 12 nM) and is supplied as a high-purity solid for laboratory research.
  • Precursor to apelin receptor agonists with reported EC50 ≈ 12 nM.
  • High purity solid (99.1%) suitable for analytical and preclinical studies.
  • Molecular formula C25H29N7O4S; molecular weight 523.61 g·mol⁻¹.
  • Available in small-scale milligram packages and as a DMSO solution for screening.
  • Intended for laboratory research and experimental use only.