Learn More
Medchemexpress LLC (2R)-4-(3-phosphonopropyl)-2-piperazinecarboxylic acid | 126453-07-4 | MFCD00153786 | 99.0% | 252.21 g/mol | C8H17N2O5P | 10 MG
SDP
Catalog No. 5000212336
Encompass_Preferred
(R)-CPP is the R-enantiomer of CPP, a potent NMDA receptor antagonist used in research to study glutamatergic signaling and NMDA receptor pharmacology. It is provided as a high-purity solid for laboratory applications and is intended for research use only.
  • Potent NMDA receptor antagonist, more active isomer (R).
  • High purity suitable for sensitive pharmacological assays.
  • Supplied as a stable solid for convenient storage and handling.
  • Suitable for in vitro and in vivo research applications.
  • Well-characterized chemical identity (CAS 126453-07-4; MW 252.21 g/mol).
Catalog No. 50-002-12336 Supplier Medchemexpress LLC Supplier No. HY10081410MG
(R)-CPP is the R-enantiomer of CPP, a potent NMDA receptor antagonist used in research to study glutamatergic signaling and NMDA receptor pharmacology. It is provided as a high-purity solid for laboratory applications and is intended for research use only.
  • Potent NMDA receptor antagonist, more active isomer (R).
  • High purity suitable for sensitive pharmacological assays.
  • Supplied as a stable solid for convenient storage and handling.
  • Suitable for in vitro and in vivo research applications.
  • Well-characterized chemical identity (CAS 126453-07-4; MW 252.21 g/mol).