Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (2R,4R)-2-(2-hydroxyphenyl)-3-(3-sulfanylpropanoyl)-1,3-thiazolidine-4-carboxylic acid | 80830-42-8 | MFCD00868438 | 99.6% | 313.39 g·mol⁻¹ | C13H15NO4S2 | 50 MG
SDP

Catalog No. 5000240770
Encompass_Preferred

Rentiapril is an orally active angiotensin-converting enzyme (ACE) inhibitor supplied for research and analytical applications. It is provided as a research reagent (not for human or clinical use) and is identified by CAS 80830-42-8. The compound has molecular formula C13H15NO4S2 and molecular weight 313.39 g·mol⁻¹.

  • Orally active ACE inhibitor with antihypertensive activity.
  • Supplied for research and analytical applications (not for human use).
  • High purity reported (99.59%).
  • Molecular weight 313.39 g·mol⁻¹.
  • Chemical formula C13H15NO4S2.
  • Available in multiple small pack sizes (5 mg-100 mg).

Catalog No. 50-002-40770 Supplier Medchemexpress LLC Supplier No. HY10644650MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Rentiapril is an orally active angiotensin-converting enzyme (ACE) inhibitor supplied for research and analytical applications. It is provided as a research reagent (not for human or clinical use) and is identified by CAS 80830-42-8. The compound has molecular formula C13H15NO4S2 and molecular weight 313.39 g·mol⁻¹.

  • Orally active ACE inhibitor with antihypertensive activity.
  • Supplied for research and analytical applications (not for human use).
  • High purity reported (99.59%).
  • Molecular weight 313.39 g·mol⁻¹.
  • Chemical formula C13H15NO4S2.
  • Available in multiple small pack sizes (5 mg-100 mg).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.