Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(2R,4R)-(-)-2,4-Pentanediol 98.0+%, TCI America™
SDP

Catalog No. P14111G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P14111G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P14111G Supplier TCI America Supplier No. P14111G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 42075-32-1
Molecular Formula C5H12O2
Molecular Weight (g/mol) 104.15
MDL Number MFCD00063893
InChI Key GTCCGKPBSJZVRZ-UHFFFAOYNA-N
Synonym 2r,4r---pentanediol, 2r,4r-pentane-2,4-diol, unii-ex4ixw1090, 2r,4r---2,4-pentanediol, 2r,4r-2,4-pentanediol, 2r,4r---2,4-dihydroxypentane, r,r---2,4-pentanediol, 2r*,4r*-2,4-pentanediol, 2,4-pentanediol #, 2,4-pentanediol,-
PubChem CID 2723683
IUPAC Name pentane-2,4-diol
SMILES CC(O)CC(C)O

Specifications

Melting Point 50°C
Color White-Yellow
Quantity 1 g
Formula Weight 104.15
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material (2R,4R)-(-)-2,4-Pentanediol
EINECSNumber (2)-0240
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.