Learn More
Enzo Life Sciences (2R,4R)-APDC (1mg). CAS: 169209-63-6
SDP
Catalog No. 502009492
Encompass_Preferred
Selective agonist for group II mGluRs. Inhibits the NADPH-oxidase dependent ROS system. Alternative name: LY-314,593, (2R,4R)-4-Aminopyrrolidine-2,4-dicarboxylic acid. Purity: ≥98% (HPLC). Solubility: Soluble in water. Long Term Storage: Ambient.
Catalog No. 50-200-9492 Supplier Enzo Life Sciences Supplier No. ALX550178M001
Selective agonist for group II mGluRs. Inhibits the NADPH-oxidase dependent ROS system. Alternative name: LY-314,593, (2R,4R)-4-Aminopyrrolidine-2,4-dicarboxylic acid. Purity: ≥98% (HPLC). Solubility: Soluble in water. Long Term Storage: Ambient.