Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (2R,4S)-5-(Biphenyl-4-yl)-4-[(tert-butoxycarbonyl)amino]-2-methylpentanoic acid | 1012341-50-2 | 100.0% | C23H29NO4 | 50 G
SDP

Catalog No. 5000434503
Encompass_Preferred

(2R,4S)-5-(Biphenyl-4-yl)-4-[(tert-butoxycarbonyl)amino]-2-methylpentanoic acid is a chemical compound primarily used as a drug intermediate in the synthesis of various active compounds. It is intended for research use only and not for human or therapeutic applications. The compound has a molecular weight of 383.48 and a high purity of 99.98%.

  • Used as a drug intermediate for the synthesis of various active compounds.
  • Intended for research use only.
  • Appears as a solid.
  • Molecular weight: 383.48.
  • Chemical formula: C23H29NO4.
  • Purity: 99.98%.
  • May cause skin irritation.
  • May cause serious eye irritation.
  • May cause respiratory irritation.
  • Harmful if swallowed.

Catalog No. 50-004-34503 Supplier Medchemexpress LLC Supplier No. HY7883750G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(2R,4S)-5-(Biphenyl-4-yl)-4-[(tert-butoxycarbonyl)amino]-2-methylpentanoic acid is a chemical compound primarily used as a drug intermediate in the synthesis of various active compounds. It is intended for research use only and not for human or therapeutic applications. The compound has a molecular weight of 383.48 and a high purity of 99.98%.

  • Used as a drug intermediate for the synthesis of various active compounds.
  • Intended for research use only.
  • Appears as a solid.
  • Molecular weight: 383.48.
  • Chemical formula: C23H29NO4.
  • Purity: 99.98%.
  • May cause skin irritation.
  • May cause serious eye irritation.
  • May cause respiratory irritation.
  • Harmful if swallowed.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.