Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (2R,4S)-5-(Biphenyl-4-yl)-4-[(tert-butoxycarbonyl)amino]-2-methylpentanoic acid | 1012341-50-2 | 99.98% | C23H29NO4 | 100 G
SDP

Catalog No. 5000434835
Encompass_Preferred

This compound is a drug intermediate used for the synthesis of various active compounds. It has a molecular weight of 383.48 and appears as a solid. This product is for research use only and not sold to patients.

  • Used as a drug intermediate for the synthesis of active compounds.
  • Molecular weight: 383.48.
  • Appearance: Solid.
  • For research use only.
  • Not sold to patients.

Catalog No. 50-004-34835 Supplier Medchemexpress LLC Supplier No. HY78837100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This compound is a drug intermediate used for the synthesis of various active compounds. It has a molecular weight of 383.48 and appears as a solid. This product is for research use only and not sold to patients.

  • Used as a drug intermediate for the synthesis of active compounds.
  • Molecular weight: 383.48.
  • Appearance: Solid.
  • For research use only.
  • Not sold to patients.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.