Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (2R,5S)-Ritlecitinib | 1792180-79-0 | MFCD29770783 | 99.8% | 285.35 g/mol | C15H19N5O | 5 MG
SDP

Supplier:  Medchemexpress LLC HY100754B5MG

Encompass_Preferred

(2R,5S)-Ritlecitinib is a selective, orally active JAK3 inhibitor supplied as a high-purity research chemical for biochemical and preclinical laboratory use. The compound is available as a solid and as a 10 mM solution in DMSO for assay preparation.

  • Selective JAK3 inhibitor for biochemical and cellular studies.
  • High purity solid suitable for analytical and preclinical work.
  • Available as 5 mg solid or as a 10 mM solution in DMSO.
  • Molecular formula C15H19N5O; molecular weight 285.35 g/mol.
  • CAS 1792180-79-0 for unambiguous substance identification.
  • Includes datasheet and SDS for handling and safety information.

Catalog No. 50-002-11663


May include imposed supplier surcharges.
Only null left
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.