Learn More
Medchemexpress LLC 2R 7aS -2-Fluorotet 1g
SDP
Catalog No. 5000205363
Encompass_Preferred
(2R 7aS)-2-Fluorotetrahydro-1H-pyrrolizine-7a(5H)-methanol is a drug intermediate for synthesis of various active compounds (2R 7aS)-2-Fluorotetrahydro-1H-pyrrolizine-7a(5H)-methanol is a core structural framework of the compounds targeting KRAS G12D mutation
Catalog No. 50-002-05363 Supplier Medchemexpress LLC Supplier No. HYW095672A1G
(2R 7aS)-2-Fluorotetrahydro-1H-pyrrolizine-7a(5H)-methanol is a drug intermediate for synthesis of various active compounds (2R 7aS)-2-Fluorotetrahydro-1H-pyrrolizine-7a(5H)-methanol is a core structural framework of the compounds targeting KRAS G12D mutation