Learn More
Medchemexpress LLC 2R-Glycerol-O-B-D-ga 1mg | 16232-91-0 | 254.23 g/mol | C9H18O8 | 1 MG
SDP
Catalog No. 5000229281
Encompass_Preferred
(2R)-Glycerol-O-β-D-galactopyranoside is a carbohydrate-derived substrate used in biochemical and enzymology studies to probe β-galactosidase activity, thiogalactoside transacetylase function, and galactoside transport systems. It is provided as a solid reagent for assay development and analytical work.
  • Substrate for β-galactosidase and thiogalactoside transacetylase.
  • Useful in studies of lac operon regulation and galactoside transport.
  • Supplied as a solid reagent suitable for small-scale assays.
  • Molecular formula C9H18O8; molecular weight ≈ 254.23 g/mol.
Catalog No. 50-002-29281 Supplier Medchemexpress LLC Supplier No. HY1160551MG
(2R)-Glycerol-O-β-D-galactopyranoside is a carbohydrate-derived substrate used in biochemical and enzymology studies to probe β-galactosidase activity, thiogalactoside transacetylase function, and galactoside transport systems. It is provided as a solid reagent for assay development and analytical work.
  • Substrate for β-galactosidase and thiogalactoside transacetylase.
  • Useful in studies of lac operon regulation and galactoside transport.
  • Supplied as a solid reagent suitable for small-scale assays.
  • Molecular formula C9H18O8; molecular weight ≈ 254.23 g/mol.