Learn More
Cayman Chemical 2R-Glycerol-OD-galactopy 1mg
SDP
Catalog No. 502965294
Encompass_Preferred
A substrate for β-galactosidase, the lactose repressor, the galactose-binding protein, the β-methylgalactoside transport system, and thiogalactoside transacetylase
Catalog No. 50-296-5294 Supplier Cayman Chemical Supplier No. 172181 MG
A substrate for β-galactosidase, the lactose repressor, the galactose-binding protein, the β-methylgalactoside transport system, and thiogalactoside transacetylase