Learn More
Medchemexpress LLC (2R)-N-[(2S)-3,3-dimethyl-1-(methylamino)-1-oxobutan-2-yl]-N'-hydroxy-2-(2-methylpropyl)butamid | 145337-55-9 | 99.4% | 315.41 g/mol | C15H29N3O4 | 100 MG
SDP
Catalog No. 5000245623
Encompass_Preferred
Ro 31-9790 is a synthetic matrix metalloproteinase (MMP) inhibitor provided for research use. It serves as a biochemical probe to study MMP-related activity in vitro and in cell-based assays. The compound is identified by CAS 145337-55-9 and is characterized by a defined molecular formula and high reported purity.
  • Acts as a matrix metalloproteinase inhibitor for research applications.
  • High purity, approximately 99.4%.
  • Molecular weight 315.41 g/mol; formula C15H29N3O4.
  • Suitable as a research-grade chemical for biochemical and cell biology studies.
  • Synonyms and systematic names are available in chemical databases for cross-referencing.
Catalog No. 50-002-45623 Supplier Medchemexpress LLC Supplier No. HY101703100MG
Ro 31-9790 is a synthetic matrix metalloproteinase (MMP) inhibitor provided for research use. It serves as a biochemical probe to study MMP-related activity in vitro and in cell-based assays. The compound is identified by CAS 145337-55-9 and is characterized by a defined molecular formula and high reported purity.
  • Acts as a matrix metalloproteinase inhibitor for research applications.
  • High purity, approximately 99.4%.
  • Molecular weight 315.41 g/mol; formula C15H29N3O4.
  • Suitable as a research-grade chemical for biochemical and cell biology studies.
  • Synonyms and systematic names are available in chemical databases for cross-referencing.