Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (2R)-octyl-α-hydroxyglutarate | 1391194-67-4 | 99.9% | 260.33 g/mol | C13H24O5 | 25 MG
SDP

Catalog No. 5000246859
Encompass_Preferred

(2R)-Octyl-α-hydroxyglutarate is the octyl ester derivative of 2-hydroxyglutarate used in biochemical and cell-based research. It is supplied as a white to off-white solid with high purity and documented solubility and storage guidelines for experimental use.

  • Solid white to off-white appearance
  • High purity (≈99.9%)
  • Molecular weight 260.33 g/mol; formula C13H24O5
  • Soluble in DMSO (100 mg/mL) and formulatable for in vivo use
  • Stable under recommended storage: 4°C sealed; in solvent -80°C (6 months)
  • Available in milligram-scale quantities including 25 mg

Catalog No. 50-002-46859 Supplier Medchemexpress LLC Supplier No. HY10364125MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(2R)-Octyl-α-hydroxyglutarate is the octyl ester derivative of 2-hydroxyglutarate used in biochemical and cell-based research. It is supplied as a white to off-white solid with high purity and documented solubility and storage guidelines for experimental use.

  • Solid white to off-white appearance
  • High purity (≈99.9%)
  • Molecular weight 260.33 g/mol; formula C13H24O5
  • Soluble in DMSO (100 mg/mL) and formulatable for in vivo use
  • Stable under recommended storage: 4°C sealed; in solvent -80°C (6 months)
  • Available in milligram-scale quantities including 25 mg

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.