Learn More
Medchemexpress LLC (2R)-octyl-α-hydroxyglutarate | 1391194-67-4 | MFCD30735809 | 99.9% | 260.33 | C13H24O5 | 10 MG
SDP
Catalog No. 5000244895
Encompass_Preferred
(2R)-Octyl-α-hydroxyglutarate is an octyl-esterified analogue of the D-isomer of 2-hydroxyglutarate. It is membrane permeable and supplied for laboratory research to probe oncometabolite biology and epigenetic regulation.
  • Membrane-permeable octyl ester of D-2-hydroxyglutarate for cellular studies.
  • Used to study oncometabolite effects on histone methylation and cell viability.
  • Reported to increase histone methylation and enhance viability of LMP1-negative NPC cells.
  • High purity (≈99.9%) suitable for biochemical experiments.
  • Available as solids (5-500 mg) and as 10 mM solution in DMSO.
  • CAS 1391194-67-4; molecular formula C13H24O5; molecular weight 260.33.
  • Store and handle per the product COA and SDS; for research use only.
Catalog No. 50-002-44895 Supplier Medchemexpress LLC Supplier No. HY10364110MG
(2R)-Octyl-α-hydroxyglutarate is an octyl-esterified analogue of the D-isomer of 2-hydroxyglutarate. It is membrane permeable and supplied for laboratory research to probe oncometabolite biology and epigenetic regulation.
  • Membrane-permeable octyl ester of D-2-hydroxyglutarate for cellular studies.
  • Used to study oncometabolite effects on histone methylation and cell viability.
  • Reported to increase histone methylation and enhance viability of LMP1-negative NPC cells.
  • High purity (≈99.9%) suitable for biochemical experiments.
  • Available as solids (5-500 mg) and as 10 mM solution in DMSO.
  • CAS 1391194-67-4; molecular formula C13H24O5; molecular weight 260.33.
  • Store and handle per the product COA and SDS; for research use only.