Learn More
Medchemexpress LLC (2R)-octyl-α-hydroxyglutarate | 1391194-67-4 | MFCD30735809 | 99.9% | 260.33 | C13H24O5 | 50 MG
SDP
Catalog No. 5000244688
Encompass_Preferred
(2R)-Octyl-α-hydroxyglutarate (CAS 1391194-67-4) is a membrane-permeable ester derivative of the oncometabolite D-2-hydroxyglutarate, provided for laboratory research use. It is used as a cell-permeable precursor to study metabolic and epigenetic effects mediated by 2-hydroxyglutarate in cultured cells.
  • Cell-permeable ester derivative of D-2-hydroxyglutarate.
  • Useful for investigating IDH mutation-related metabolic pathways.
  • Supplied at high purity suitable for biochemical and cellular assays.
  • Available in small milligram pack sizes for laboratory experiments.
  • For research use only; not for human or clinical use.
Catalog No. 50-002-44688 Supplier Medchemexpress LLC Supplier No. HY10364150MG
Add to Cart
Add to Cart
(2R)-Octyl-α-hydroxyglutarate (CAS 1391194-67-4) is a membrane-permeable ester derivative of the oncometabolite D-2-hydroxyglutarate, provided for laboratory research use. It is used as a cell-permeable precursor to study metabolic and epigenetic effects mediated by 2-hydroxyglutarate in cultured cells.
  • Cell-permeable ester derivative of D-2-hydroxyglutarate.
  • Useful for investigating IDH mutation-related metabolic pathways.
  • Supplied at high purity suitable for biochemical and cellular assays.
  • Available in small milligram pack sizes for laboratory experiments.
  • For research use only; not for human or clinical use.