Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (2R)-Octyl-α-hydroxyglutarate | 1391194-67-4 | MFCD30735809 | 99.9% | 260.33 g/mol | C13H24O5 | 1 ML
SDP

Catalog No. 5000245041
Encompass_Preferred

(2R)-Octyl-α-hydroxyglutarate is a cell-permeable octyl-esterified form of the D-isomer 2-hydroxyglutarate used in biochemical and cell biology research to model 2-HG activity and investigate metabolic and epigenetic effects. It is provided as a ready-to-use solution for cell treatment and as solid material for assay and analytical work.

  • Molecular formula: C13H24O5; molecular weight: 260.33 g/mol.
  • High purity: 99.91% (as reported).
  • Cell-permeable octyl ester for efficient intracellular delivery of 2-HG activity.
  • Common applications: metabolic studies, epigenetic research, and cell-based assays.
  • Available as a 10 mM solution in DMSO (1 mL) and in multiple solid sizes for analytical use.
  • Storage: sealed at 4°C; in solution, store at -80°C for long-term stability.

Catalog No. 50-002-45041 Supplier Medchemexpress LLC Supplier No. HY10364110MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(2R)-Octyl-α-hydroxyglutarate is a cell-permeable octyl-esterified form of the D-isomer 2-hydroxyglutarate used in biochemical and cell biology research to model 2-HG activity and investigate metabolic and epigenetic effects. It is provided as a ready-to-use solution for cell treatment and as solid material for assay and analytical work.

  • Molecular formula: C13H24O5; molecular weight: 260.33 g/mol.
  • High purity: 99.91% (as reported).
  • Cell-permeable octyl ester for efficient intracellular delivery of 2-HG activity.
  • Common applications: metabolic studies, epigenetic research, and cell-based assays.
  • Available as a 10 mM solution in DMSO (1 mL) and in multiple solid sizes for analytical use.
  • Storage: sealed at 4°C; in solution, store at -80°C for long-term stability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.