Learn More
Medchemexpress LLC (2R)-octyl-α-hydroxyglutarate | 1391194-67-4 | MFCD30735809 | 99.9% | 260.33 g·mol⁻¹ | C13H24O5 | 100 MG
SDP
Catalog No. 5000244586
Encompass_Preferred
(2R)-Octyl-α-hydroxyglutarate is a cell-permeable, esterified derivative of D-2-hydroxyglutarate used as a research reagent to investigate the biological effects of 2-HG in cellular systems, including effects on histone methylation, DNA damage, and mitochondrial oxidative processes.
  • Cell-permeable analogue of D-2-hydroxyglutarate.
  • Used to study histone methylation and epigenetic regulation.
  • Reported purity approximately 99.9%.
  • Molecular weight 260.33 g·mol⁻¹; chemical formula C13H24O5.
  • Provided as an analytical/research standard in small pack sizes suitable for cell-based assays.
Catalog No. 50-002-44586 Supplier Medchemexpress LLC Supplier No. HY103641100MG
Add to Cart
Add to Cart
(2R)-Octyl-α-hydroxyglutarate is a cell-permeable, esterified derivative of D-2-hydroxyglutarate used as a research reagent to investigate the biological effects of 2-HG in cellular systems, including effects on histone methylation, DNA damage, and mitochondrial oxidative processes.
  • Cell-permeable analogue of D-2-hydroxyglutarate.
  • Used to study histone methylation and epigenetic regulation.
  • Reported purity approximately 99.9%.
  • Molecular weight 260.33 g·mol⁻¹; chemical formula C13H24O5.
  • Provided as an analytical/research standard in small pack sizes suitable for cell-based assays.