Learn More
Cayman Chemical (2R)-Octyl--hydroxyglutarate, 1391194-67-4, 5 mg
SDP
Supplier:  Cayman Chemical 163665
Encompass_Preferred
Molecular Formula C13H24O5, Purity ≥95%, Formula Weight 260.3, Synonyms: 2R-hydroxy-pentanedioic acid, 1-octyl ester, (2R)-Octyl-2-HG; A cell-permeable derivative of the D-isomer of 2-HG; used to examine the contribution of D-2HG to the oxidative mitochondrial processes of IDH1-mutated cancer cells.
Catalog No. NC1027882