Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STA Pharmaceutical (2R,3R)/(2S,3S)-Racemic Fmoc-beta-Me-Phenylalanine | 5 g | CAS 321524-79-2 | MDL MFCD06656477

Catalog No. 502342489
Encompass

(2R,3R)/(2S,3S)-Racemic Fmoc-beta-Me-Phenylalanine is a Amino Acid reagent (Subcategory: Phe) sold by WuXi TIDES. Offered in 5 g. Store at 4 °C. SDS available for reference.

Specifications
- CAS: 321524-79-2
- MDL: MFCD06656477
- InChIKey: YDULLWGMRWMFLU-FZKOXMLESA-N
- Molecular Weight: 802.924
- Molecular Formula: C50H46N2O8
- Purity: ≥95%
- Container Type: 30 mL HDPE
- Pack Size: 5 g
- Net Weight: 5 g
- Gross Weight: 16.5 g
- Commodity Code: 29242970
- Country Of Origin: China
- IUPAC: (2R,3R)-2-((((9H-fluoren-9-yl)methoxy)carbonyl)amino)-3-phenylbutanoic acid--(2S,3S)-2-((((9H-fluoren-9-yl)methoxy)carbonyl)amino)-3-phenylbutanoic acid (1/1)
- SMILES: C[C@H]([C@@H](C(O)=O)NC(OCC1C2=CC=CC=C2C3=CC=CC=C31)=O)C4=CC=CC=C4.C[C@@H]([C@H](C(O)=O)NC(OCC5C6=CC=CC=C6C7=CC=CC=C75)=O)C8=CC=CC=C8

Catalog No. 50-234-2489 Supplier STA Pharmaceutical Supplier No. STAAA120205GM
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(2R,3R)/(2S,3S)-Racemic Fmoc-beta-Me-Phenylalanine is a Amino Acid reagent (Subcategory: Phe) sold by WuXi TIDES. Offered in 5 g. Store at 4 °C. SDS available for reference.

Specifications
- CAS: 321524-79-2
- MDL: MFCD06656477
- InChIKey: YDULLWGMRWMFLU-FZKOXMLESA-N
- Molecular Weight: 802.924
- Molecular Formula: C50H46N2O8
- Purity: ≥95%
- Container Type: 30 mL HDPE
- Pack Size: 5 g
- Net Weight: 5 g
- Gross Weight: 16.5 g
- Commodity Code: 29242970
- Country Of Origin: China
- IUPAC: (2R,3R)-2-((((9H-fluoren-9-yl)methoxy)carbonyl)amino)-3-phenylbutanoic acid--(2S,3S)-2-((((9H-fluoren-9-yl)methoxy)carbonyl)amino)-3-phenylbutanoic acid (1/1)
- SMILES: C[C@H]([C@@H](C(O)=O)NC(OCC1C2=CC=CC=C2C3=CC=CC=C31)=O)C4=CC=CC=C4.C[C@@H]([C@H](C(O)=O)NC(OCC5C6=CC=CC=C6C7=CC=CC=C75)=O)C8=CC=CC=C8

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.