Learn More
Cayman Chemical (2R)-Octyl-α-hydroxyglutarate, Purity: ≥95%, CAS: 1391194-67-4, 10 mg
SDP
Catalog No. 502971055
Encompass_Preferred
A cell-permeable derivative of the D-isomer of 2-HG; used to examine the contribution of D-2HG to the oxidative mitochondrial processes of IDH1-mutated cancer cells
Catalog No. 50-297-1055 Supplier Cayman Chemical Supplier No. 1636610 MG
A cell-permeable derivative of the D-isomer of 2-HG; used to examine the contribution of D-2HG to the oxidative mitochondrial processes of IDH1-mutated cancer cells