Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(2RS)-2-(4-Formylphenyl)propionic acid Pharmaceutical Impurity Standard, ≥98.0% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111014194 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-4194 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-4194 Supplier MilliporeSigma Supelco Supplier No. 7989550MG
Add to Cart
Add to Cart

Specifications

CAS 43153-07-7
Linear Formula C10H10O3
MDL Number MFCD08275588
Synonym (2 RS)-2-(4-Formylphenyl)propanoic acid
Type Pharmaceutical Impurity Standard
Quantity 50 mg
Formula Weight 178.18
Percent Purity ≥98.0% (HPLC)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.