Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (2S)-1-[(tert-butoxy)carbonyl]-2-methylazetidine-2-carboxylic acid | 1363402-35-0 | MFCD22395149 | 97.1% | 215.25 g/mol | C10H17NO4 | 100 MG
SDP

Catalog No. 5000210530
Encompass_Preferred

(2S)-1-[(tert-butoxy)carbonyl]-2-methylazetidine-2-carboxylic acid is a Boc-protected azetidine carboxylic acid used as a drug intermediate in medicinal chemistry and organic synthesis. It is intended for research and synthetic applications, and is supplied with analytical data supporting its stated purity.

  • Purity: 97.1%.
  • Molecular weight: 215.25 g/mol.
  • Chemical formula: C10H17NO4.
  • CAS number: 1363402-35-0.
  • Available in small-scale packaging for research use (100 mg-10 g).
  • Function: synthetic building block and intermediate for substituted azetidine compounds.

Catalog No. 50-002-10530 Supplier Medchemexpress LLC Supplier No. HYW063820100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(2S)-1-[(tert-butoxy)carbonyl]-2-methylazetidine-2-carboxylic acid is a Boc-protected azetidine carboxylic acid used as a drug intermediate in medicinal chemistry and organic synthesis. It is intended for research and synthetic applications, and is supplied with analytical data supporting its stated purity.

  • Purity: 97.1%.
  • Molecular weight: 215.25 g/mol.
  • Chemical formula: C10H17NO4.
  • CAS number: 1363402-35-0.
  • Available in small-scale packaging for research use (100 mg-10 g).
  • Function: synthetic building block and intermediate for substituted azetidine compounds.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.