Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (2S)-1-[(tert-butoxy)carbonyl]-2-methylazetidine-2-carboxylic acid | 1363402-35-0 | MFCD22395149 | 97.1% | 215.25 g/mol | C10H17NO4 | 250 MG
SDP

Catalog No. 5000209689
Encompass_Preferred

(2S)-1-[(tert-Butoxy)carbonyl]-2-methylazetidine-2-carboxylic acid is a Boc-protected azetidine carboxylic acid used as a building block and drug intermediate in medicinal chemistry synthesis. It is supplied as a solid with specified storage conditions and analytical purity suitable for research applications.

  • Used as a building block and drug intermediate in medicinal chemistry
  • High purity: 97.1%
  • Molecular weight: 215.25 g/mol
  • Chemical formula: C10H17NO4
  • Available in multiple package sizes; pack shown: 250 MG
  • Storage: powder -20°C (3 years), 4°C (2 years); in solvent -80°C (6 months), -20°C (1 month)

Catalog No. 50-002-09689 Supplier Medchemexpress LLC Supplier No. 250MGHYW063820
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(2S)-1-[(tert-Butoxy)carbonyl]-2-methylazetidine-2-carboxylic acid is a Boc-protected azetidine carboxylic acid used as a building block and drug intermediate in medicinal chemistry synthesis. It is supplied as a solid with specified storage conditions and analytical purity suitable for research applications.

  • Used as a building block and drug intermediate in medicinal chemistry
  • High purity: 97.1%
  • Molecular weight: 215.25 g/mol
  • Chemical formula: C10H17NO4
  • Available in multiple package sizes; pack shown: 250 MG
  • Storage: powder -20°C (3 years), 4°C (2 years); in solvent -80°C (6 months), -20°C (1 month)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.