Learn More
Strem, An Ascensus Company CAS# 1490388-03-8. 50mg. (2S)-2-[[[[3,5-Bis(trifluoromethyl)phenyl]amino]thioxomethyl]amino]-3,3-dimethyl-N-(phenylmethyl)butanamide, 98%, (99% ee)
SDP
Supplier:  Strem, An Ascensus Company 07063150MG
Encompass
CAS# 1490388-03-8. 50mg. (2S)-2-[[[[3,5-Bis(trifluoromethyl)phenyl]amino]thioxomethyl]amino]-3,3-dimethyl-N-(phenylmethyl)butanamide, 98%, (99% ee). Molecular Weight: 491.50. Molecular Formula: C22H23F6N3OS. Color/form: white to yellow pwdr. Strem# 07-0631. http://www.strem.com/catalog/v/07-0631/
Catalog No. NC2085007