Learn More
Medchemexpress LLC (2S)-2-[[3-cyclobutyl-5-(3,4,5-trifluorophenoxy)imidazo[4,5-b]pyridin-2-yl]methylamino]propanamide | 1233231-30-5 | 98.3% | 419.4 g/mol | C20H20F3N5O2 | 100MG
SDP
Catalog No. 5000235776
Encompass_Preferred
DSP-2230 (ANP-230) is an orally active small-molecule inhibitor of voltage-gated sodium channels with activity against Nav1.7, Nav1.8, and Nav1.9. It is commonly used in preclinical research on neuropathic pain and sodium channel pharmacology, and is supplied at research-grade purity for in vitro and in vivo studies.
  • Inhibits Nav1.7, Nav1.8, and Nav1.9 with reported IC50s of 7.1 μM, 11.4 μM, and 6.7 μM respectively.
  • High purity suitable for research use (≈98.3%).
  • Small-molecule structure with molecular weight 419.4 g/mol.
  • Soluble in DMSO for assay preparation.
  • Available in multiple pack sizes to support dose-ranging and scaling studies.
Catalog No. 50-002-35776 Supplier Medchemexpress LLC Supplier No. HY125079100MG
DSP-2230 (ANP-230) is an orally active small-molecule inhibitor of voltage-gated sodium channels with activity against Nav1.7, Nav1.8, and Nav1.9. It is commonly used in preclinical research on neuropathic pain and sodium channel pharmacology, and is supplied at research-grade purity for in vitro and in vivo studies.
  • Inhibits Nav1.7, Nav1.8, and Nav1.9 with reported IC50s of 7.1 μM, 11.4 μM, and 6.7 μM respectively.
  • High purity suitable for research use (≈98.3%).
  • Small-molecule structure with molecular weight 419.4 g/mol.
  • Soluble in DMSO for assay preparation.
  • Available in multiple pack sizes to support dose-ranging and scaling studies.