Learn More
Medchemexpress LLC (2S)-2-[[3-cyclobutyl-5-(3,4,5-trifluorophenoxy)imidazo[4,5-b]pyridin-2-yl]methylamino]propanamide | 1233231-30-5 | 98.3% | 419.4 g/mol | C20H20F3N5O2 | 10MG
SDP
Catalog No. 5000238269
Encompass_Preferred
DSP-2230 (ANP-230) is an orally active small-molecule inhibitor of voltage-gated sodium channels, developed for research into neuropathic pain and sodium-channel pharmacology. It is used in both in vitro and in vivo studies to evaluate inhibition of Nav1.7, Nav1.8, and Nav1.9 and to probe analgesic mechanisms. The compound is supplied as a high-purity research reagent with good solubility in DMSO.
Catalog No. 50-002-38269 Supplier Medchemexpress LLC Supplier No. HY12507910MG
DSP-2230 (ANP-230) is an orally active small-molecule inhibitor of voltage-gated sodium channels, developed for research into neuropathic pain and sodium-channel pharmacology. It is used in both in vitro and in vivo studies to evaluate inhibition of Nav1.7, Nav1.8, and Nav1.9 and to probe analgesic mechanisms. The compound is supplied as a high-purity research reagent with good solubility in DMSO.