Learn More
Medchemexpress LLC (2S)-2-[[3-cyclobutyl-5-(3,4,5-trifluorophenoxy)imidazo[4,5-b]pyridin-2-yl]methylamino]propanamide | 1233231-30-5 | 98.3% | 419.4 g/mol | C20H20F3N5O2 | 25MG
SDP
Catalog No. 5000237969
Encompass_Preferred
DSP-2230 (ANP-230) is an orally active voltage-gated sodium channel inhibitor used in preclinical research to modulate Nav1.7, Nav1.8, and Nav1.9 activity and to evaluate approaches for neuropathic pain. The compound is supplied as a solid with characterized solubility profiles and storage recommendations for laboratory use only.
  • Inhibits Nav1.7, Nav1.8, and Nav1.9 with reported IC50s of 7.1 μM, 11.4 μM, and 6.7 μM respectively.
  • Reported purity 98.33%.
  • Soluble in DMSO at ≥100 mg/mL; multiple in vivo formulation protocols with solubility ≥2.25 mg/mL are provided.
  • Powder stability: -20°C for up to 3 years; solution stability conditions are reported.
  • Intended for preclinical studies of sodium channel pharmacology and neuropathic pain models.
Catalog No. 50-002-37969 Supplier Medchemexpress LLC Supplier No. HY12507925MG
DSP-2230 (ANP-230) is an orally active voltage-gated sodium channel inhibitor used in preclinical research to modulate Nav1.7, Nav1.8, and Nav1.9 activity and to evaluate approaches for neuropathic pain. The compound is supplied as a solid with characterized solubility profiles and storage recommendations for laboratory use only.
  • Inhibits Nav1.7, Nav1.8, and Nav1.9 with reported IC50s of 7.1 μM, 11.4 μM, and 6.7 μM respectively.
  • Reported purity 98.33%.
  • Soluble in DMSO at ≥100 mg/mL; multiple in vivo formulation protocols with solubility ≥2.25 mg/mL are provided.
  • Powder stability: -20°C for up to 3 years; solution stability conditions are reported.
  • Intended for preclinical studies of sodium channel pharmacology and neuropathic pain models.