Learn More
Medchemexpress LLC (2S)-2-[[3-cyclobutyl-5-(3,4,5-trifluorophenoxy)imidazo[4,5-b]pyridin-2-yl]methylamino]propanamide | 1233231-30-5 | 98.3% | 419.4 g/mol | C20H20F3N5O2 | 50MG
SDP
Catalog No. 5000238140
Encompass_Preferred
DSP-2230 (ANP-230) is an orally active inhibitor of voltage-gated sodium channels with activity against Nav1.7, Nav1.8, and Nav1.9. It is used in preclinical research to evaluate analgesic effects and sodium channel modulation in models of neuropathic pain. The compound is supplied as a solid with high purity and documented solubility and storage recommendations for both in vitro and in vivo applications.
  • Orally active inhibitor of Nav1.7, Nav1.8, and Nav1.9.
  • Suitable for in vitro and in vivo studies, with DMSO solubility ≥100 mg/mL.
  • High purity suitable for research use.
  • Supplied as a solid powder with recommended cold storage to maintain stability.
  • Documented molecular weight and formula support analytical characterization.
Catalog No. 50-002-38140 Supplier Medchemexpress LLC Supplier No. HY12507950MG
DSP-2230 (ANP-230) is an orally active inhibitor of voltage-gated sodium channels with activity against Nav1.7, Nav1.8, and Nav1.9. It is used in preclinical research to evaluate analgesic effects and sodium channel modulation in models of neuropathic pain. The compound is supplied as a solid with high purity and documented solubility and storage recommendations for both in vitro and in vivo applications.
  • Orally active inhibitor of Nav1.7, Nav1.8, and Nav1.9.
  • Suitable for in vitro and in vivo studies, with DMSO solubility ≥100 mg/mL.
  • High purity suitable for research use.
  • Supplied as a solid powder with recommended cold storage to maintain stability.
  • Documented molecular weight and formula support analytical characterization.