Learn More
Medchemexpress LLC [(2S)-2-(4-chlorophenyl)-1-pyrrolidinyl](3-methyl-1H-pyrazolo[3,4-b]pyridin-5-yl)-methanone | 1883423-59-3 | 99.6% | 340.81 | C18H17ClN4O | 1 MG
SDP
Catalog No. 5000249250
Encompass_Preferred
MSC2530818 is a potent, selective, orally available cyclin-dependent kinase 8 (CDK8) inhibitor supplied as a research-grade analytical standard for biochemical and cellular studies. It is intended for use in in vitro kinase assays, cellular potency testing, and as an analytical reference material; high purity and characterization enable reproducible results.
  • Potent and selective CDK8 inhibition (IC50 = 2.6 nM).
  • High purity (≈99.6%) suitable for analytical applications.
  • Available as a solid and as a 10 mM solution in DMSO for convenience.
  • Fully characterized with molecular weight 340.81 and formula C18H17ClN4O.
  • Provided with supporting documentation (COA and SDS) for traceability.
Catalog No. 50-002-49250 Supplier Medchemexpress LLC Supplier No. HY101611R1MG
MSC2530818 is a potent, selective, orally available cyclin-dependent kinase 8 (CDK8) inhibitor supplied as a research-grade analytical standard for biochemical and cellular studies. It is intended for use in in vitro kinase assays, cellular potency testing, and as an analytical reference material; high purity and characterization enable reproducible results.
  • Potent and selective CDK8 inhibition (IC50 = 2.6 nM).
  • High purity (≈99.6%) suitable for analytical applications.
  • Available as a solid and as a 10 mM solution in DMSO for convenience.
  • Fully characterized with molecular weight 340.81 and formula C18H17ClN4O.
  • Provided with supporting documentation (COA and SDS) for traceability.