Learn More
Medchemexpress LLC (2S)-2-(4-fluorophenyl)-1-[(4-methylphenyl)sulfonyl]-pyrrolidine | 298690-60-5 | MFCD20270617 | 99.7% | 319.39 | C17H18FNO2S | 2 MG
SDP
Catalog No. 5000214124
Encompass_Preferred
Ro 67-7476 is a selective positive allosteric modulator (PAM) of the metabotropic glutamate receptor 1 (mGluR1). It potentiates glutamate-induced calcium release and activates ERK1/2 signaling in cellular assays; supplied for research use only.
  • Selective positive allosteric modulator of mGluR1.
  • Potentiates glutamate-induced calcium release (EC50 = 60.1 nM).
  • Activates ERK1/2 phosphorylation in cell-based assays (EC50 = 163.3 nM).
  • High reported purity suitable for biochemical and pharmacology studies.
  • Available as solid and as DMSO solution for assay flexibility.
Catalog No. 50-002-14124 Supplier Medchemexpress LLC Supplier No. HY1004032MG
Ro 67-7476 is a selective positive allosteric modulator (PAM) of the metabotropic glutamate receptor 1 (mGluR1). It potentiates glutamate-induced calcium release and activates ERK1/2 signaling in cellular assays; supplied for research use only.
  • Selective positive allosteric modulator of mGluR1.
  • Potentiates glutamate-induced calcium release (EC50 = 60.1 nM).
  • Activates ERK1/2 phosphorylation in cell-based assays (EC50 = 163.3 nM).
  • High reported purity suitable for biochemical and pharmacology studies.
  • Available as solid and as DMSO solution for assay flexibility.