Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (2S)-2-amino-3-[(3-fluorophenyl)-(2-phenylmethoxyphenyl)methoxy]propanoic acid; TFA | 00-00-0 | 99.8% | 509.45 g·mol⁻¹ | C25H23F4NO6 | 5 MG
SDP

Catalog No. 5000263712
Encompass_Preferred

ALX-1393 TFA is the trifluoroacetate salt of a selective glycine transporter 2 (GlyT2) inhibitor used as a research reagent to investigate glycinergic neurotransmission and nociception. It is supplied as a purified solid for laboratory use and has reported antinociceptive activity in rodent pain models.

  • Selective GlyT2 inhibitor for probing glycinergic neurotransmission.
  • Research-grade solid suitable for in vitro and in vivo studies.
  • High purity (99.83%) supporting reproducible results.
  • Molecular formula C25H23F4NO6 and molecular weight 509.45 g·mol⁻¹.
  • Store at -20°C to maintain stability and minimize moisture exposure.

Catalog No. 50-002-63712 Supplier Medchemexpress LLC Supplier No. HY111029A5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ALX-1393 TFA is the trifluoroacetate salt of a selective glycine transporter 2 (GlyT2) inhibitor used as a research reagent to investigate glycinergic neurotransmission and nociception. It is supplied as a purified solid for laboratory use and has reported antinociceptive activity in rodent pain models.

  • Selective GlyT2 inhibitor for probing glycinergic neurotransmission.
  • Research-grade solid suitable for in vitro and in vivo studies.
  • High purity (99.83%) supporting reproducible results.
  • Molecular formula C25H23F4NO6 and molecular weight 509.45 g·mol⁻¹.
  • Store at -20°C to maintain stability and minimize moisture exposure.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.