Learn More
Medchemexpress LLC (2S)-2-[(tert-Butoxycarbonyl)amino]-3-(3,4-difluorophenyl)propionic acid | 198474-90-7 | 98.0% | C14H17F2NO4 | 10 G

Supplier: Medchemexpress LLC HY7916510G
(2S)-2-[(tert-Butoxycarbonyl)amino]-3-(3,4-difluorophenyl)propionic acid is a solid laboratory chemical intended for research use. It is stable under recommended storage conditions and has a molecular weight of 301.29.
- Solid appearance
- Melting/freezing point: 95-105°C
- Boiling point: 430.4°C at 760 mmHg
- Flash point: 214.1°C
- Relative density: 1.27 g/cm³
- Stable under recommended storage conditions
- May cause respiratory irritation
- Causes skin irritation
- Causes serious eye irritation
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.