Learn More
Medchemexpress LLC (2S)-2-[(tert-Butoxycarbonyl)amino]-3-(3,4-difluorophenyl)propionic acid | 198474-90-7 | 98.0% | C14H17F2NO4 | 5 G
SDP
Catalog No. 5000435036
Encompass_Preferred
(2S)-2-[(tert-Butoxycarbonyl)amino]-3-(3,4-difluorophenyl)propionic acid is a phenylalanine derivative. Amino acids and amino acid derivatives have been commercially used as ergogenic supplements, influencing anabolic hormone secretion, fuel supply during exercise, mental performance during stress-related tasks, and preventing exercise-induced muscle damage. They are recognized as beneficial ergogenic dietary substances.
  • Solid appearance
  • White to light yellow color
  • Ships at room temperature in continental US
  • Powder can be stored at -20°C for 3 years or 4°C for 2 years
  • In solvent, store at -80°C for 6 months or -20°C for 1 month
  • Soluble in DMSO at ≥ 100 mg/mL (331.91 mM)
  • For research use only
Catalog No. 50-004-35036 Supplier Medchemexpress LLC Supplier No. HY791655G
(2S)-2-[(tert-Butoxycarbonyl)amino]-3-(3,4-difluorophenyl)propionic acid is a phenylalanine derivative. Amino acids and amino acid derivatives have been commercially used as ergogenic supplements, influencing anabolic hormone secretion, fuel supply during exercise, mental performance during stress-related tasks, and preventing exercise-induced muscle damage. They are recognized as beneficial ergogenic dietary substances.
  • Solid appearance
  • White to light yellow color
  • Ships at room temperature in continental US
  • Powder can be stored at -20°C for 3 years or 4°C for 2 years
  • In solvent, store at -80°C for 6 months or -20°C for 1 month
  • Soluble in DMSO at ≥ 100 mg/mL (331.91 mM)
  • For research use only