Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(2S,3aS,7aS)-1-Fmoc-octahydroindole-2-carboxylic acid, 98%

Catalog No. AAH6349103
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6349103 1 g
AAH63491MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6349103 Supplier Thermo Scientific Chemicals Supplier No. H6349103
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 130309-37-4 | C24H25NO4 | 391.467 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 130309-37-4
Molecular Formula C24H25NO4
Molecular Weight (g/mol) 391.467
MDL Number MFCD00191201
InChI Key JBZXLQHJZHITMW-RXYZOABWSA-N
Synonym fmoc-oic-oh, 2s,3as,7as-1-9h-fluoren-9-yl methoxy carbonyl octahydro-1h-indole-2-carboxylic acid, fmoc-2s,3as,7as-octahydro-1h-indole-2-carboxylic acid, 2s,3as,7as-1-9h-fluoren-9-ylmethoxy carbonyl-octahydroindole-2-carboxylic acid, fmoc-l-octahydroindole-2-carboxylic acid, ambotzfaa1729, 2s pound not3as pound not7as-1-9h-fluoren-9-yl methoxy carbonyl, 2s,3as,7as-1-9h-fluoren-9-yl methoxy carbonyl-octahydro-1h-indole-2-carboxylic acid, 2s,3as,7as-1-9h-fluoren-9-ylmethoxycarbonyl-2,3,3a,4,5,6,7,7a-octahydroindole-2-carboxylic acid
PubChem CID 2756165
IUPAC Name (2S,3aS,7aS)-1-(9H-fluoren-9-ylmethoxycarbonyl)-2,3,3a,4,5,6,7,7a-octahydroindole-2-carboxylic acid
SMILES C1CCC2C(C1)CC(N2C(=O)OCC3C4=CC=CC=C4C5=CC=CC=C35)C(=O)O

Specifications

Color White
Quantity 1 g
Formula Weight 391.46
Percent Purity 98%
Physical Form Powder
Chemical Name or Material (2S,3aS,7aS)-1-Fmoc-octahydroindole-2-carboxylic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.