Learn More
Medchemexpress LLC (2S,3R)-2,3,4-Trihydroxybutanal-13C-2 | 90913-09-0 | 99.4% | 121.10 | 5 MG
SDP
Catalog No. 5000454941
Encompass_Preferred
(2S,3R)-2,3,4-Trihydroxybutanal-13C-2 is a 13C labeled compound primarily used in research. It incorporates stable heavy isotopes for quantitation in drug development processes.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Incorporates stable heavy isotopes of carbon for quantitation.
Catalog No. 50-004-54941 Supplier Medchemexpress LLC Supplier No. HYW012864S25MG
(2S,3R)-2,3,4-Trihydroxybutanal-13C-2 is a 13C labeled compound primarily used in research. It incorporates stable heavy isotopes for quantitation in drug development processes.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Incorporates stable heavy isotopes of carbon for quantitation.