Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (2S,3R)-2,3,4-Trihydroxybutanal-13C-2 | 90913-09-0 | 99.4% | 121.10 | 50 MG
SDP

Catalog No. 5000454979
Encompass_Preferred

(2S,3R)-2,3,4-Trihydroxybutanal-13C-2 is the 13C labeled (2S,3R)-2,3,4-Trihydroxybutanal.

  • Used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process.

Catalog No. 50-004-54979 Supplier Medchemexpress LLC Supplier No. HYW012864S250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(2S,3R)-2,3,4-Trihydroxybutanal-13C-2 is the 13C labeled (2S,3R)-2,3,4-Trihydroxybutanal.

  • Used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.