Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(2S,3R)-2,3-Dihydroxybutyric acid sodium salt hydrate ≥97.0% (GC), MilliporeSigma™ Supelco™

Catalog No. 111003804 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-100-3804 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-100-3804 Supplier MilliporeSigma Supelco Supplier No. 9094850MG

Specifications

Linear Formula C4H8O4 · xNa+ · yH2 O
Synonym (2 S,3 R)-2,3-Dihydroxybutanoic acid sodium salt hydrate; 4-Deoxy-D-threonic acid sodium salt hydrate
Type Organic Standard
Quantity 50 mg
Formula Weight 120.10 (anhydrous free acid basis)
Percent Purity ≥97% (GC)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.