Learn More
Bachem [(2S,3R)-3-Amino-2-hydroxy-4-phenylbutyryl]-L-leucine
SDP
Supplier:  Bachem 4006587.0050
Encompass_Preferred
[(2S,3R)-3-Amino-2-hydroxy-4-phenylbutyryl]-L-leucine, 58970-76-6, C16H24N2O4, Mr 308.38, 50mg. Bestatin, Ubenimex. Bestatin (or ubenimex), an antibiotic of natural origin, is a specific, competitive inhibitor of many aminopeptidases, though it does not inhibit aminopeptidase A. Due to its low toxicity it has been administered to cultured cells, animals and humans. Bestatin offered by Bachem has been obtained by chemical synthesis.
Catalog No. 50-259-119