Learn More
Apexbio Technology LLC 2S-3R-4S-5R Amino TH Hexanal
SDP
Catalog No. 5000897557
Encompass_Preferred
(2S 3R 4S 5R)-2-Amino-3 4 5 6-tetrahydroxyhexanal hydrochloride (CAS No 5505-63-5) is a compound of interest in the biomedical research category of carbohydrate chemistry Originating from monosaccharide derivatives it acts primarily by interacting with pathways involved in glucose metabolism and cellular signaling In vitro this compound exhibits activity typically in the nanomolar to low micromolar range making it suitable for detailed biochemical assays It is commonly utilized in cell models to study glycosylation processes and occasionally in animal studies to explore metabolic disorders Additionally it is applied in drug screening to identify potential inhibitors of carbohydrate-metabolizing enzymes Experimental concentrations typically depend on the specific research design This compound s versatility in research applications underscores its importance as a tool for understanding biological processes related to carbohydrates
Catalog No. 50-008-97557 Supplier Apexbio Technology LLC Supplier No. C6547200
(2S 3R 4S 5R)-2-Amino-3 4 5 6-tetrahydroxyhexanal hydrochloride (CAS No 5505-63-5) is a compound of interest in the biomedical research category of carbohydrate chemistry Originating from monosaccharide derivatives it acts primarily by interacting with pathways involved in glucose metabolism and cellular signaling In vitro this compound exhibits activity typically in the nanomolar to low micromolar range making it suitable for detailed biochemical assays It is commonly utilized in cell models to study glycosylation processes and occasionally in animal studies to explore metabolic disorders Additionally it is applied in drug screening to identify potential inhibitors of carbohydrate-metabolizing enzymes Experimental concentrations typically depend on the specific research design This compound s versatility in research applications underscores its importance as a tool for understanding biological processes related to carbohydrates