Learn More
Medchemexpress LLC HY-12422B 5mg Medchemexpress, (2S,3R)-Voruciclib (hydrochloride) CAS: Purity:>98%
SDP
Catalog No. 501969661
Encompass_Preferred
Medchemexpress, HY-12422B 5mg (2S,3R)-Voruciclib (HCl) CAS: (2S,3R)-Voruciclib hydrochloride is the less active enantiomer of Voruciclib hydrochloride. Voruciclib is a clinical stage orally active and selective CDK inhibitor with Ki values of 0.626 nM-9.1 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9661 Supplier Medchemexpress LLC Supplier No. HY12422B5MG
Add to Cart
Add to Cart
Medchemexpress, HY-12422B 5mg (2S,3R)-Voruciclib (HCl) CAS: (2S,3R)-Voruciclib hydrochloride is the less active enantiomer of Voruciclib hydrochloride. Voruciclib is a clinical stage orally active and selective CDK inhibitor with Ki values of 0.626 nM-9.1 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.